Gene Transcripts

Gene Symbol
Gas7
Entrez Gene ID
85246
Gene Name
growth arrest specific 7
Gene Aliases
Chromosome Location
Chr. 10: 54010722 - 54240798 on Build Rnor_6.0
UniGene ID
Rn.17160
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053484.2 NP_445936.2 7 548
XM_006246827.2 XP_006246889.1 7 1217
XM_006246828.2 XP_006246890.1 7 2058
XM_006246829.2 XP_006246891.1 7 1401
XM_006246830.2 XP_006246892.1 7 732
XM_008767883.1 XP_008766105.1 7 1132
GenBank mRNA AJ003148.1 - 7 684
AJ131902.1 - 684

View Full Product Details