Gene Transcripts

Gene Symbol
TAF5L
Entrez Gene ID
27097
Gene Name
TATA-box binding protein associated factor 5 like
Gene Aliases
PAF65B
Chromosome Location
Chr. 1: 229593111 - 229626047 on Build GRCh38
UniGene ID
Hs.270621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025247.1 NP_001020418.1 4 615
NM_014409.3 NP_055224.1 4 615
XM_005273099.4 XP_005273156.1 4 568
XM_005273100.2 XP_005273157.1 5 581
XM_011544162.1 XP_011542464.1 3 352
XM_011544163.1 XP_011542465.1 7 1197
XM_011544164.2 XP_011542466.1 3 295
XM_017001045.1 XP_016856534.1 6 1352
GenBank mRNA AF069736.1 - 3 449
BC041094.1 - 4 538 5551246

View Full Product Details