Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
29666
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Psmb6l
Chromosome Location
Chr. 10: 57131347 - 57133685 on Build Rnor_6.0
UniGene ID
Rn.8118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057099.3 NP_476440.2 4 367
XM_008767811.1 XP_008766033.1 4 398
GenBank mRNA BC058451.1 - 4 371
D10754.1 - 4 356
FQ209811.1 - 4 370
FQ217385.1 - 4 370
FQ224639.1 - 4 370

Gene Symbol
LOC100360846
Entrez Gene ID
100360846
Gene Name
proteasome subunit beta type 6-like
Gene Aliases
Chromosome Location
Chr. 2: 96031948 - 96033565 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729028.3 XP_002729074.2 2 515

View Full Product Details