Gene Transcripts

Gene Symbol
Kcnc3
Entrez Gene ID
117101
Gene Name
potassium channel, voltage gated Shaw-related subfamily C, member 3
Gene Aliases
KShIIID, Kv3.3
Chromosome Location
Chr. 1: 100593476 - 100607874 on Build Rnor_6.0
UniGene ID
Rn.9885
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053997.4 NP_446449.2 2 2189
XM_006228965.2 XP_006229027.1 2 2383
XM_006228967.2 XP_006229029.1 2 2383
XM_006228971.2 XP_006229033.1 2 2383
XM_006228972.2 XP_006229034.1 2 2383
XM_006228973.2 XP_006229035.1 2 2383
XM_008759306.1 XP_008757528.1 2 2383
XM_008759307.1 XP_008757529.1 2 2383
XM_008759308.1 XP_008757530.1 2 2383
XM_008759309.1 XP_008757531.1 2 2383
GenBank mRNA AY179603.1 - 2 2189
AY179604.1 - 2 2189
AY822673.1 - 2 2189
AY822674.1 - 2 2189
M84210.1 - 2 2189
M84211.2 - 2 2189

View Full Product Details