Gene Transcripts

Gene Symbol
HERC5
Entrez Gene ID
51191
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Gene Aliases
CEB1, CEBP1
Chromosome Location
Chr. 4: 88456604 - 88506170 on Build GRCh38
UniGene ID
Hs.26663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016323.3 NP_057407.2 2 474
XM_011532022.2 XP_011530324.2 2 987
GenBank mRNA AB027289.1 - 2 434
AY337518.1 - 2 474
BC140716.1 - 2 367 9021584

View Full Product Details