Gene Transcripts

Gene Symbol
Fgf19
Entrez Gene ID
170582
Gene Name
fibroblast growth factor 19
Gene Aliases
Fgf15
Chromosome Location
Chr. 1: 218058405 - 218061693 on Build Rnor_6.0
UniGene ID
Rn.81230
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130753.1 NP_570109.1 1 123
GenBank mRNA AB078900.1 - 1 123

View Full Product Details