Gene Transcripts

Gene Symbol
Ap3m2
Entrez Gene ID
140667
Gene Name
adaptor-related protein complex 3, mu 2 subunit
Gene Aliases
Chromosome Location
Chr. 16: 74075842 - 74091230 on Build Rnor_6.0
UniGene ID
Rn.11007
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133305.2 NP_579839.1 3 549
GenBank mRNA BC086993.1 - 3 549
L07074.1 - 372

View Full Product Details