Gene Transcripts

Gene Symbol
Ick
Entrez Gene ID
84411
Gene Name
intestinal cell kinase
Gene Aliases
Chromosome Location
Chr. 8: 85413975 - 85473374 on Build Rnor_6.0
UniGene ID
Rn.3750
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138886.1 NP_620241.1 3, 4 454
XM_008766345.1 XP_008764567.1 3, 4 677
GenBank mRNA D26178.1 - 3, 4 454

View Full Product Details