Gene Transcripts

Gene Symbol
Ick
Entrez Gene ID
84411
Gene Name
intestinal cell kinase
Gene Aliases
Chromosome Location
Chr. 8: 85413975 - 85473374 on Build Rnor_6.0
UniGene ID
Rn.3750
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138886.1 NP_620241.1 12 1828
XM_008766345.1 XP_008764567.1 12 2051
GenBank mRNA D26178.1 - 12 1828

View Full Product Details