Gene Transcripts

Gene Symbol
Klhl12
Entrez Gene ID
266772
Gene Name
kelch-like family member 12
Gene Aliases
C3ip1
Chromosome Location
Chr. 13: 51280933 - 51314753 on Build Rnor_6.0
UniGene ID
Rn.15513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153730.2 NP_714952.1 2 247
XM_006249843.2 XP_006249905.1 3 396
XM_006249844.2 XP_006249906.1 3 670
XM_006249845.1 XP_006249907.1 3 376
XM_006249846.2 XP_006249908.1 4 462
XM_008769539.1 XP_008767761.1 3 435
XM_008769540.1 XP_008767762.1 4 290
XM_008769541.1 XP_008767763.1 3 180
GenBank mRNA AJ133126.1 - 2 289
BC086983.1 - 3 377 7133597
FQ210179.1 - 4 446

View Full Product Details