Gene Transcripts

Gene Symbol
Npas4
Entrez Gene ID
266734
Gene Name
neuronal PAS domain protein 4
Gene Aliases
Nxf
Chromosome Location
Chr. 1: 220260310 - 220278024 on Build Rnor_6.0
UniGene ID
Rn.86654
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153626.1 NP_705890.1 7 1501
XM_006230697.2 XP_006230759.1 9 3658
GenBank mRNA AB050103.1 - 7 1501

View Full Product Details