Gene Transcripts

Gene Symbol
Aip
Entrez Gene ID
282827
Gene Name
aryl-hydrocarbon receptor-interacting protein
Gene Aliases
XAP2
Chromosome Location
Chr. 1: 219361859 - 219373912 on Build Rnor_6.0
UniGene ID
Rn.95160
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172327.2 NP_758830.2 4 544
XM_006230699.2 XP_006230761.1 5 742
XM_006230700.2 XP_006230762.1 5 711
GenBank mRNA AF543560.1 - 4 578
BC089110.1 - 4 544 7311852
FQ214270.1 - 4 652
FQ228146.1 - 4 572

View Full Product Details