Gene Transcripts

Gene Symbol
Tmem37
Entrez Gene ID
245953
Gene Name
transmembrane protein 37
Gene Aliases
Pr, Pr1
Chromosome Location
Chr. 13: 36094665 - 36101440 on Build Rnor_6.0
UniGene ID
Rn.92180
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139095.1 NP_620795.1 2 217
XM_008769424.1 XP_008767646.1 2 577
GenBank mRNA AF361355.1 - 2 217

View Full Product Details