Gene Transcripts

Gene Symbol
Rab11b
Entrez Gene ID
79434
Gene Name
RAB11B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 7: 18598991 - 18612132 on Build Rnor_6.0
UniGene ID
Rn.124832
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032617.2 NP_116006.1 2 156
GenBank mRNA AF286534.1 - 2 162
BC062041.1 - 2 156 5598168
D01046.1 - 2 142

View Full Product Details