Gene Transcripts

Gene Symbol
Csn1s2a
Entrez Gene ID
12993
Gene Name
casein alpha s2-like A
Gene Aliases
Csng, PP22
Chromosome Location
Chr. 5: 87774567 - 87788797 on Build GRCm38
UniGene ID
Mm.474059
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK142688.1 - 3, 4 137
AK164738.1 - 3, 4 144
BC139326.1 - 3, 4 117 8862348
D10215.1 - 3, 4 142

View Full Product Details