Gene Transcripts

Gene Symbol
Ddx19a
Entrez Gene ID
13680
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a
Gene Aliases
DBP5, Ddx19, Eif4a-rs1
Chromosome Location
Chr. 8: 110974556 - 110997823 on Build GRCm38
UniGene ID
Mm.287901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007916.2 NP_031942.2 12 2376
GenBank mRNA AK146548.1 - 12 2337
BC011270.1 - 12 2340 3674608
L25125.1 - 12 2368

View Full Product Details