Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008062.2 | NP_032088.1 | 13 | 2130 | |
GenBank mRNA | AK088135.1 | - | 13 | 1851 | |
AK148031.1 | - | 13 | 1853 | ||
AK165207.1 | - | 13 | 1884 | ||
BC075663.1 | - | 13 | 2130 | 30603864 | |
Z11911.1 | - | 13 | 1835 |