Gene Transcripts

Gene Symbol
Gdf3
Entrez Gene ID
14562
Gene Name
growth differentiation factor 3
Gene Aliases
C78318, Gdf-3, Vgr-2, Vgr2, ecat9
Chromosome Location
Chr. 6: 122605403 - 122610071 on Build GRCm38
UniGene ID
Mm.299742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008108.4 NP_032134.2 1, 2 383
GenBank mRNA AK082877.1 - 1, 2 304
AK146174.1 - 1, 2 309
AK152457.1 - 1, 2 384
AK160533.1 - 1, 2 341
AK167962.1 - 1, 2 309
BC101963.1 - 1, 2 298 40045099
BC101964.1 - 1, 2 298 40045100
BC103565.1 - 1, 2 297 40045101
L06443.1 - 1, 2 399
S52658.1 - 1, 2 339

View Full Product Details