Gene Transcripts

Gene Symbol
Kcnd1
Entrez Gene ID
16506
Gene Name
potassium voltage-gated channel, Shal-related family, member 1
Gene Aliases
1110037K09Rik, Kca2-1, Kv4.1, Shal, Shal1, mShal1
Chromosome Location
Chr. X: 7823843 - 7836503 on Build GRCm38
UniGene ID
Mm.335968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC139325.1 - 1 212
BC139327.1 - 1 212 8862349
M64226.1 - 1 128

View Full Product Details