Gene Transcripts

Gene Symbol
Il9
Entrez Gene ID
16198
Gene Name
interleukin 9
Gene Aliases
Il-9, P40
Chromosome Location
Chr. 13: 56479277 - 56482246 on Build GRCm38
UniGene ID
Mm.3006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008373.1 NP_032399.1 4 304
GenBank mRNA X14045.1 - 4 304

View Full Product Details