Gene Transcripts
- Gene Symbol
- Nap1l4
- Entrez Gene ID
- 17955
- Gene Name
- nucleosome assembly protein 1-like 4
- Gene Aliases
- 2810410H14Rik, AI316776, D7Wsu30e, Nap2
- Chromosome Location
- Chr. 7: 143513579 - 143549196 on Build GRCm38
- UniGene ID
- Mm.294625
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285489.1 |
NP_001272418.1
|
12, 13 |
1127 |
|
NM_001285490.1 |
NP_001272419.1
|
12, 13 |
1140 |
|
NM_008672.3 |
NP_032698.1
|
12, 13 |
1127 |
|
GenBank mRNA |
AJ002198.1 |
- |
12, 13 |
1097 |
|
AK090112.1 |
- |
12, 13 |
1143 |
|
AK156835.1 |
- |
12, 13 |
1100 |
|
BC089018.1 |
- |
12, 13 |
1114 |
|
BC132039.1 |
- |
11, 12 |
1051 |
40130316 |
BC138518.1 |
- |
11, 12 |
1051 |
8861538 |
BC145306.1 |
- |
11, 12 |
1051 |
9053860 |
BC145308.1 |
- |
11, 12 |
1051 |
9053862 |
View Full Product Details