Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
19175
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Lmp19, Mpnd
Chromosome Location
Chr. 11: 70525357 - 70527858 on Build GRCm38
UniGene ID
Mm.98
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008946.4 NP_032972.3 4 399
GenBank mRNA AK078437.1 - 4 371
AK132042.1 - 4 382
AK167123.1 - 4 371
AK167227.1 - 4 367
BC013897.1 - 4 359 4038758
U13393.1 - 4 320

View Full Product Details