Gene Transcripts
- Gene Symbol
- Exoc4
- Entrez Gene ID
- 20336
- Gene Name
- exocyst complex component 4
- Gene Aliases
- C78892, Sec8, Sec8l1
- Chromosome Location
- Chr. 6: 33249090 - 33972943 on Build GRCm38
- UniGene ID
- Mm.265512
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009148.3 |
NP_033174.2
|
1, 2 |
97 |
|
GenBank mRNA |
AF022962.1 |
- |
1, 2 |
123 |
|
AK010695.1 |
- |
1, 2 |
112 |
|
AK014499.1 |
- |
1, 2 |
120 |
|
AK036507.1 |
- |
1, 2 |
109 |
|
AK038125.1 |
- |
1, 2 |
120 |
|
AK039983.1 |
- |
1, 2 |
120 |
|
AK086603.1 |
- |
1, 2 |
116 |
|
AK173235.1 |
- |
1, 2 |
100 |
|
BC034644.1 |
- |
1, 2 |
97 |
5355276 |
View Full Product Details