Gene Transcripts

Gene Symbol
Tbx2
Entrez Gene ID
21385
Gene Name
T-box 2
Gene Aliases
Chromosome Location
Chr. 11: 85832615 - 85841948 on Build GRCm38
UniGene ID
Mm.287052
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009324.2 NP_033350.2 2 945
GenBank mRNA AK080703.1 - 1 587
AK137225.1 - 2 151
AK149348.1 - 2 753
U15566.1 - 2 481

View Full Product Details