Gene Transcripts

Gene Symbol
Umod
Entrez Gene ID
22242
Gene Name
uromodulin
Gene Aliases
THP, Urehd1, urehr4
Chromosome Location
Chr. 7: 119462708 - 119479262 on Build GRCm38
UniGene ID
Mm.10826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278605.1 NP_001265534.1 3 433
NM_009470.5 NP_033496.1 3 489
GenBank mRNA AK085460.1 - 3 483
AK144065.1 - 3 405
AK164688.1 - 3 405
AK165507.1 - 3 349
BC012973.1 - 2 327 4210940
L33406.1 - 3 419

View Full Product Details