Gene Transcripts

Gene Symbol
Ckmt1
Entrez Gene ID
12716
Gene Name
creatine kinase, mitochondrial 1, ubiquitous
Gene Aliases
Mt-CK, ScCKmit, UbCKmit, mi-CK
Chromosome Location
Chr. 2: 121357751 - 121363737 on Build GRCm38
UniGene ID
Mm.252145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009897.2 NP_034027.1 2, 3 568
GenBank mRNA AK005076.1 - 2, 3 567
AK018487.1 - 2, 3 551
AK131953.1 - 2, 3 572
BC025976.1 - 3, 4 483 4216758
Z13968.1 - 2, 3 568

View Full Product Details