Gene Transcripts

Gene Symbol
Csf2ra
Entrez Gene ID
12982
Gene Name
colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage)
Gene Aliases
CD116, Csfgmra, GM-CSF-Ra, GM-CSFR
Chromosome Location
Chr. 19: 61224401 - 61228465 on Build GRCm38
UniGene ID
Mm.287228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009970.2 NP_034100.2 12 1449
GenBank mRNA AK135114.1 - 5 1431
M85078.1 - 12 1390

View Full Product Details