Gene Transcripts

Gene Symbol
Fut9
Entrez Gene ID
14348
Gene Name
fucosyltransferase 9
Gene Aliases
AI746471, AU067636, mFUT9, mFuc-TIX
Chromosome Location
Chr. 4: 25609333 - 25800003 on Build GRCm38
UniGene ID
Mm.39101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010243.3 NP_034373.1 3 786
GenBank mRNA AB015426.1 - 3 390
AK032177.1 - 3 928
AK033607.1 - 2 941
AK036183.1 - 3 589
AK043410.1 - 3 627
AK047650.1 - 3 585
AK049122.1 - 4 867
AK163871.1 - 3 586
BC116874.1 - 3 482 40126193
BC116876.1 - 3 482 40126195

View Full Product Details