Gene Transcripts

Gene Symbol
Fut9
Entrez Gene ID
14348
Gene Name
fucosyltransferase 9
Gene Aliases
AI746471, AU067636, mFUT9, mFuc-TIX
Chromosome Location
Chr. 4: 25609333 - 25800003 on Build GRCm38
UniGene ID
Mm.39101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010243.3 NP_034373.1 3 3335
GenBank mRNA AK047650.1 - 3 3134
AK163871.1 - 3 3135

View Full Product Details