Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039129.4 | NP_001034218.1 | 2 | 217 | |
NM_010447.5 | NP_034577.1 | 2 | 217 | ||
GenBank mRNA | AK007802.1 | - | 2 | 163 | |
AK088308.1 | - | 2 | 162 | ||
AK131999.1 | - | 2 | 183 | ||
AK135391.1 | - | 2 | 161 | ||
AK136113.1 | - | 2 | 185 | ||
AK146126.1 | - | 2 | 161 | ||
AK146630.1 | - | 2 | 183 | ||
AK152684.1 | - | 2 | 161 | ||
AK161730.1 | - | 2 | 161 | ||
AK166901.1 | - | 2 | 161 | ||
AK166989.1 | - | 2 | 159 | ||
AK167161.1 | - | 2 | 161 | ||
AK167810.1 | - | 2 | 161 | ||
AK167811.1 | - | 2 | 161 | ||
AK167820.1 | - | 2 | 163 | ||
AK167913.1 | - | 2 | 161 | ||
AK168420.1 | - | 2 | 161 | ||
AK168531.1 | - | 2 | 164 | ||
AK168733.1 | - | 2 | 161 | ||
AK169216.1 | - | 2 | 161 | ||
BC052723.1 | - | 2 | 147 | ||
BC080675.1 | - | 2 | 143 | 6308746 | |
BC083136.1 | - | 2 | 128 | 6511966 | |
BC089340.1 | - | 2 | 136 | 6402927 | |
BC092395.1 | - | 2 | 137 | 6403376 | |
BY067931.1 | - | 2 | 219 | ||
D86728.1 | - | 2 | 124 | ||
D86729.1 | - | 2 | 124 | ||
M99167.1 | - | 2 | 159 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001165971.1 | NP_001159443.1 | 1 | 254 | |
GenBank mRNA | BC157911.1 | - | 1 | 254 | |
BC172092.1 | - | 1 | 254 |