Gene Transcripts

Gene Symbol
Ifit3
Entrez Gene ID
15959
Gene Name
interferon-induced protein with tetratricopeptide repeats 3
Gene Aliases
Ifi49, P49
Chromosome Location
Chr. 19: 34583142 - 34588982 on Build GRCm38
UniGene ID
Mm.426079
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010501.2 NP_034631.1 2 229
GenBank mRNA AK077459.1 - 2 227
AK151032.1 - 2 229
AK153226.1 - 2 230
BC003804.1 - 2 195 3487345
BC089563.1 - 2 227 30317428
CT010266.1 - 1 158
L32974.1 - 2 210

Gene Symbol
I830012O16Rik
Entrez Gene ID
667370
Gene Name
RIKEN cDNA I830012O16 gene
Gene Aliases
Ifit3b
Chromosome Location
Chr. 19: 34607539 - 34613409 on Build GRCm38
UniGene ID
Mm.271850
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005858.3 NP_001005858.2 195
GenBank mRNA AK077243.1 - 184
CA576017.1 - 195

View Full Product Details