Gene Transcripts

Gene Symbol
Il17a
Entrez Gene ID
16171
Gene Name
interleukin 17A
Gene Aliases
Ctla-8, Ctla8, IL-17, IL-17A, Il17
Chromosome Location
Chr. 1: 20730905 - 20734496 on Build GRCm38
UniGene ID
Mm.5419
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010552.3 NP_034682.1 2 209
GenBank mRNA AK040420.1 - 2 210
BC119303.1 - 2 209 8734052
BC119309.1 - 2 209 8734058
U43088.1 - 2 196

View Full Product Details