Gene Transcripts
- Gene Symbol
- Pcbp2
- Entrez Gene ID
- 18521
- Gene Name
- poly(rC) binding protein 2
- Gene Aliases
- AW412548, Hnrpx, alphaCP-2
- Chromosome Location
- Chr. 15: 102470222 - 102500059 on Build GRCm38
- UniGene ID
- Mm.236513
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001103165.1 |
NP_001096635.1
|
2, 3 |
322 |
|
NM_001103166.1 |
NP_001096636.1
|
2, 3 |
322 |
|
NM_001174073.1 |
NP_001167544.1
|
2, 3 |
298 |
|
NM_011042.2 |
NP_035172.2
|
2, 3 |
322 |
|
GenBank mRNA |
AK012297.1 |
- |
2, 3 |
170 |
|
AK161527.1 |
- |
2, 3 |
146 |
|
AK169268.1 |
- |
2, 3 |
416 |
|
BC020478.1 |
- |
1, 2 |
67 |
|
BQ900329.1 |
- |
2, 3 |
298 |
|
EU604548.1 |
- |
1, 2 |
77 |
|
L19661.1 |
- |
2, 3 |
209 |
|
View Full Product Details