Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
18752
Gene Name
protein kinase C, gamma
Gene Aliases
PKCgamma, Pkcc, Prkcc
Chromosome Location
Chr. 7: 3303532 - 3331099 on Build GRCm38
UniGene ID
Mm.7980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291434.1 NP_001278363.1 1 362
NM_011102.4 NP_035232.1 1 362
GenBank mRNA AK144419.1 - 1 364
BC111807.1 - 1 249 40039740
L28035.1 - 1 440
X67129.1 - 1 171

View Full Product Details