Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011192.3 | NP_035322.1 | 11 | 1868 | |
GenBank mRNA | AK140597.1 | - | 11 | 1890 | |
AK182317.1 | - | 1 | 372 | ||
AK219671.1 | - | 1 | 274 | ||
BC005524.1 | - | 3 | 1121 | ||
BC015255.1 | - | 11 | 1868 | 4221225 | |
BC139818.1 | - | 11 | 1816 | 40141268 | |
D87911.1 | - | 11 | 1791 |