Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011237.2 | NP_035367.1 | 11 | 1640 | |
GenBank mRNA | AF045663.1 | - | 11 | 1640 | |
AK138973.1 | - | 10 | 1718 | ||
AK165833.1 | - | 11 | 1676 | ||
BC082556.1 | - | 11 | 1579 | 30538342 |