Gene Transcripts

Gene Symbol
Tcea1
Entrez Gene ID
21399
Gene Name
transcription elongation factor A (SII) 1
Gene Aliases
S-II
Chromosome Location
Chr. 1: 4857694 - 4897909 on Build GRCm38
UniGene ID
Mm.207263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159750.1 NP_001153222.1 7 845
NM_001159751.1 NP_001153223.1 7 741
NM_011541.4 NP_035671.1 7 848
GenBank mRNA AK136035.1 - 7 741
AK143593.1 - 7 706
BC006022.1 - 7 723 3590733
BC061490.1 - 7 740 2582185
BC083127.1 - 7 711 6390364
M18209.1 - 6 644
M18210.1 - 7 702

View Full Product Details