Gene Transcripts

Gene Symbol
Slc30a3
Entrez Gene ID
22784
Gene Name
solute carrier family 30 (zinc transporter), member 3
Gene Aliases
Znt3
Chromosome Location
Chr. 5: 31086106 - 31108244 on Build GRCm38
UniGene ID
Mm.1396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011773.3 NP_035903.2 5 836
GenBank mRNA AK144568.1 - 5 820
AK165796.1 - 5 799
BC066199.1 - 5 818 30476359
U76007.1 - 5 794

View Full Product Details