Gene Transcripts

Gene Symbol
Grem2
Entrez Gene ID
23893
Gene Name
gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis)
Gene Aliases
Gremlin2, Prdc
Chromosome Location
Chr. 1: 174833785 - 174921915 on Build GRCm38
UniGene ID
Mm.25760
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011825.1 NP_035955.1 2 361
GenBank mRNA AB011030.1 - 2 361
AK142008.1 - 2 363
AK161827.1 - 2 341
BC079905.1 - 2 342 30645267

View Full Product Details