Gene Transcripts

Gene Symbol
Lynx1
Entrez Gene ID
23936
Gene Name
Ly6/neurotoxin 1
Gene Aliases
AI838844, SLURP-2
Chromosome Location
Chr. 15: 74747856 - 74752979 on Build GRCm38
UniGene ID
Mm.257067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011838.4 NP_035968.1 4 411
GenBank mRNA AF141377.1 - 4 385
AK013827.1 - 4 408
AK161907.1 - 4 441
AK162920.1 - 4 434
AK165295.1 - 4 415
AK165314.1 - 4 415
BC037541.1 - 4 458 5325123

View Full Product Details