Gene Transcripts

Gene Symbol
Nlrp5
Entrez Gene ID
23968
Gene Name
NLR family, pyrin domain containing 5
Gene Aliases
Mater, Nalp5, Op1, PAN11
Chromosome Location
Chr. 7: 23385897 - 23441921 on Build GRCm38
UniGene ID
Mm.333653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF074018.1 - 3, 4 229
AK087774.1 - 4, 5 297
AY196361.1 - 3, 4 229
AY196362.1 - 3, 4 229
AY329484.1 - 3, 4 229
AY329485.1 - 3, 4 229
AY329486.1 - 3, 4 229
AY329487.1 - 3, 4 229
AY329488.1 - 3, 4 229
AY329489.1 - 3, 4 229
AY329490.1 - 3, 4 229
AY329491.1 - 3, 4 229
BC053384.1 - 4, 5 296 30049556

View Full Product Details