Gene Transcripts

Gene Symbol
Mmel1
Entrez Gene ID
27390
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
Mell1, NEP2, NEPII, Nl1, SEP
Chromosome Location
Chr. 4: 154869585 - 154895530 on Build GRCm38
UniGene ID
Mm.116944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013783.2 NP_038811.2 4 539
GenBank mRNA AF157105.1 - 4 485
AF157106.1 - 2 194
AF176569.1 - 4 536
AF302075.1 - 2 160
AF302076.1 - 3 229
AF302077.1 - 2 160
AK154366.1 - 1 28
BC137649.1 - 2 312 8860668
BC144804.1 - 2 312 9053352

View Full Product Details