Gene Transcripts

Gene Symbol
Hs6st1
Entrez Gene ID
50785
Gene Name
heparan sulfate 6-O-sulfotransferase 1
Gene Aliases
6Ost1
Chromosome Location
Chr. 1: 36068400 - 36106446 on Build GRCm38
UniGene ID
Mm.213566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015818.2 NP_056633.2 2 1322
GenBank mRNA AB024566.1 - 2 1313
AK144809.1 - 3 1242
AK157556.1 - 2 1186
AK165161.1 - 1 2362
AK165385.1 - 2 725
AK166962.1 - 427
AK167151.1 - 2 751
BC052316.1 - 2 1322 6391204

View Full Product Details