Gene Transcripts
- Gene Symbol
- Sufu
- Entrez Gene ID
- 24069
- Gene Name
- suppressor of fused homolog (Drosophila)
- Gene Aliases
- Su(fu)
- Chromosome Location
- Chr. 19: 46396896 - 46488804 on Build GRCm38
- UniGene ID
- Mm.41210
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025391.2 |
NP_001020562.1
|
11, 12 |
1577 |
|
NM_015752.3 |
NP_056567.2
|
11, 12 |
1580 |
|
GenBank mRNA |
AF134893.1 |
- |
11, 12 |
1520 |
|
AJ131692.1 |
- |
11, 12 |
1578 |
|
AJ308626.1 |
- |
11, 12 |
1511 |
|
AK016976.1 |
- |
11, 12 |
1558 |
|
AK156410.1 |
- |
11, 12 |
1542 |
|
AK196809.1 |
- |
2, 3 |
152 |
|
AK200545.1 |
- |
2, 3 |
152 |
|
BC048168.1 |
- |
11, 12 |
1550 |
6821477 |
BC056997.1 |
- |
11, 12 |
1550 |
6821477 |
View Full Product Details