Gene Transcripts

Gene Symbol
Stip1
Entrez Gene ID
20867
Gene Name
stress-induced phosphoprotein 1
Gene Aliases
Hop, Sti1, p60
Chromosome Location
Chr. 19: 7020696 - 7040026 on Build GRCm38
UniGene ID
Mm.258633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016737.2 NP_058017.1 2, 3 365
GenBank mRNA AK075988.1 - 2, 3 303
AK088494.1 - 2, 3 307
AK134171.1 - 2, 3 281
AK149493.1 - 2, 3 281
AK161645.1 - 2, 3 284
AK167273.1 - 2, 3 299
AK168181.1 - 2, 3 281
AK181280.1 - 2, 3 238
CJ155828.1 - 2, 3 366
U27830.1 - 2, 3 282

View Full Product Details