Gene Transcripts
- Gene Symbol
- Mybph
- Entrez Gene ID
- 53311
- Gene Name
- myosin binding protein H
- Gene Aliases
- AI385643, MyBP-H
- Chromosome Location
- Chr. 1: 134193417 - 134201234 on Build GRCm38
- UniGene ID
- Mm.379067
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016749.2 |
NP_058029.2
|
|
1824 |
|
GenBank mRNA |
U68267.1 |
- |
|
1818 |
|
View Full Product Details