Gene Transcripts

Gene Symbol
Dgka
Entrez Gene ID
13139
Gene Name
diacylglycerol kinase, alpha
Gene Aliases
80kDa, AW146112, Dagk1
Chromosome Location
Chr. 10: 128720134 - 128748420 on Build GRCm38
UniGene ID
Mm.291235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016811.2 NP_058091.2 3 418
GenBank mRNA AF085219.1 - 2 104
BC006713.1 - 3 418 3584592
JN832677.1 - 4 450
JN832678.1 - 3 437
JN832679.1 - 3 437

View Full Product Details