Gene Transcripts

Gene Symbol
Ybx2
Entrez Gene ID
53422
Gene Name
Y box protein 2
Gene Aliases
Msy2
Chromosome Location
Chr. 11: 69935835 - 69941604 on Build GRCm38
UniGene ID
Mm.29286
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016875.2 NP_058571.2 3 438
GenBank mRNA AF073954.1 - 3 365
AF073955.1 - 3 232
AK131786.1 - 3 416
BC100454.1 - 3 438
BC141112.1 - 3 541 9055724
DV661016.1 - 2 40

View Full Product Details