Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017371.2 | NP_059067.2 | 4 | 300 | |
GenBank mRNA | AK145928.1 | - | 4 | 298 | |
BC011246.1 | - | 4 | 280 | 4196684 | |
BC019901.1 | - | 4 | 282 | 5123914 | |
BY088352.1 | - | 4 | 301 | ||
U89889.1 | - | 4 | 256 |