Gene Transcripts

Gene Symbol
Ostf1
Entrez Gene ID
20409
Gene Name
osteoclast stimulating factor 1
Gene Aliases
C78236, SH3P2, Sh3d3
Chromosome Location
Chr. 19: 18580364 - 18631813 on Build GRCm38
UniGene ID
Mm.172222
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017375.3 NP_059071.1 10 1155
GenBank mRNA AK002899.1 - 10 1134
AK005525.1 - 10 1124
AK008467.1 - 10 1103
AK010074.1 - 10 1151
AK145949.1 - 10 1106
AK149175.1 - 1133
AK150145.1 - 10 1125
AK205236.1 - 1 215
AK209921.1 - 1 295
BC060986.1 - 10 1034 30300544

View Full Product Details